Anti-IgA/IgG/IgM Goat Polyclonal Antibody (PE (Phycoerythrin)/Texas Red®)

Lieferant: Abcam
AB130774-125
ABCAAB130774-125EA 412 EUR
ABCAAB130774-125
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (PE (Phycoerythrin)/Texas Red®)
Antikörper
Type: Secondary
Antigen: Mouse IgG+IgM+IgA
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR