Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin)/Texas Red®)

Lieferant: Abcam
AB130801-125
ABCAAB130801-125EA 463 EUR
ABCAAB130801-125
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin)/Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR