Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: DR-4-02]

Lieferant: Abcam
AB14738-100
ABCAAB14738-100EA 1410 EUR
ABCAAB14738-100
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: DR-4-02]
Antikörper
Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: monoclonal
Clone: DR-4-02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR