Anti-IgM Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Abcam
AB150189-500
ABCAAB150189-500EA 269 EUR
ABCAAB150189-500
Anti-IgM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Rabbit Anti-Human IgM mu chain (Alexa Fluor® 488).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR