Anti-Placental lactogen Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB15555-7
ABCAAB15555-7EA 858 EUR
ABCAAB15555-7
Anti-Placental lactogen Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Placental lactogen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR