Anti-EMR1 Rat Monoclonal Antibody Biotin [clone: BM8]

Lieferant: Abcam
AB15694-50
ABCAAB15694-50EA 398 EUR
ABCAAB15694-50
Anti-EMR1 Rat Monoclonal Antibody Biotin [clone: BM8]
Antikörper
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: Monoclonal
Clone: BM8
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR