Anti-MYCN Mouse Monoclonal Antibody [clone: NCM II 100]
Lieferant: Abcam
AB16898-50
ABCAAB16898-50EA 862 EUR
ABCAAB16898-50
Anti-MYCN Mouse Monoclonal Antibody [clone: NCM II 100]
Antikörper
Type: Primary
Antigen: MYCN (v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog)
Clonality: Monoclonal
Clone: NCM II 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse