Anti-BCL2 Mouse Monoclonal Antibody [clone: 10C4]

Lieferant: Abcam
AB16904
ABCAAB16904EA 1000 EUR
ABCAAB16904
Anti-BCL2 Mouse Monoclonal Antibody [clone: 10C4]
Antikörper
Type: Primary
Antigen: BCL2 (B-cell CLL/lymphoma 2)
Clonality: monoclonal
Clone: 10C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR