Anti-Cleaved PARP1 Mouse Monoclonal Antibody [clone: 4B5BD2] (Alexa Fluor® 488)

Lieferant: Abcam
AB170171-100
ABCAAB170171-100EA 732 EUR
ABCAAB170171-100
Anti-Cleaved PARP1 Mouse Monoclonal Antibody [clone: 4B5BD2] (Alexa Fluor® 488)
Antikörper
Alexa Fluor® 488 Mouse monoclonal [4B5BD2] to Cleaved PARP1.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cleaved PARP1
Clonality: Monoclonal
Clone: 4B5BD2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR