Anti-Histone H3 (di methyl K9) Rabbit Monoclonal Antibody [clone: EP16990]

Lieferant: Abcam
AB176882-100 AB176882-1ML
ABCAAB176882-100EA 700 EUR
ABCAAB176882-100 ABCAAB176882-1ML
Anti-Histone H3 (di methyl K9) Rabbit Monoclonal Antibody [clone: EP16990]
Antikörper
Rabbit monoclonal [EP16990] to Histone H3 (di methyl K9) - ChIP Grade.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EP16990
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR