Anti-Mitochondria Mouse Monoclonal Antibody [clone: SPM198]

Lieferant: Abcam
AB17857-7
ABCAAB17857-7EA 918 EUR
ABCAAB17857-7
Anti-Mitochondria Mouse Monoclonal Antibody [clone: SPM198]
Antikörper
Type: Primary
Antigen: Mitochondria
Clonality: monoclonal
Clone: SPM198
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR