Anti-SYP Rabbit Monoclonal Antibody [clone: YE269] (Alexa Fluor® 488)

Lieferant: Abcam
AB196379-100
ABCAAB196379-100EA 757 EUR
ABCAAB196379-100
Anti-SYP Rabbit Monoclonal Antibody [clone: YE269] (Alexa Fluor® 488)
Antikörper
Alexa Fluor® 488 Rabbit monoclonal [YE269] to Synaptophysin.

Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: YE269
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR