Anti-Lamin B2 Monoclonal Antibody [clone: EPR9701(B)] (Alexa Fluor® 647)

Lieferant: Abcam
AB200427-100
ABCAAB200427-100EA 807 EUR
ABCAAB200427-100
Anti-Lamin B2 Monoclonal Antibody [clone: EPR9701(B)] (Alexa Fluor® 647)
Antikörper
Alexa Fluor® 647 Rabbit monoclonal [EPR9701(B)] to Lamin B2.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Lamin B2
Clonality: Monoclonal
Clone: EPR9701(B)
Conjugation: Alexa Fluor® 647
Epitope:
Host:
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR