Anti-HLA DR beta Mouse Monoclonal Antibody [clone: DA2]

Lieferant: Abcam
AB20184-50
ABCAAB20184-50EA 870 EUR
ABCAAB20184-50
Anti-HLA DR beta Mouse Monoclonal Antibody [clone: DA2]
Antikörper
Type: Primary
Antigen: HLA DR beta
Clonality: monoclonal
Clone: DA2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR