Anti-HOXB4 Mouse Monoclonal Antibody [clone: 3A2F2]
Lieferant: Abcam
AB201957-100
ABCAAB201957-100EA 700 EUR
ABCAAB201957-100
Anti-HOXB4 Mouse Monoclonal Antibody [clone: 3A2F2]
Antikörper
Mouse monoclonal [3A2F2] to HOXB4.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HOXB4
Clonality: Monoclonal
Clone: 3A2F2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human