Anti-Hsp90 beta Rabbit Monoclonal Antibody [clone: EPR16621] (Alexa Fluor® 488)

Lieferant: Abcam
AB202822-100
ABCAAB202822-100EA 825 EUR
ABCAAB202822-100
Anti-Hsp90 beta Rabbit Monoclonal Antibody [clone: EPR16621] (Alexa Fluor® 488)
Antikörper
Alexa Fluor® 488 Rabbit monoclonal [EPR16621] to Hsp90 beta.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Hsp90 beta
Clonality: Monoclonal
Clone: EPR16621
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR