Anti-Histone H3 Mouse Monoclonal Antibody [clone: mAbcam 1220] (Alexa Fluor® 488)

Lieferant: Abcam
AB203850-100
ABCAAB203850-100EA 707 EUR
ABCAAB203850-100
Anti-Histone H3 Mouse Monoclonal Antibody [clone: mAbcam 1220] (Alexa Fluor® 488)
Antikörper
Alexa Fluor® 488 Mouse monoclonal [mAbcam 1220] to Histone H3 (di methyl K9).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: mAbcam 1220
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR