Anti-Anillin Mouse Monoclonal Antibody [clone: CL0303]

Lieferant: Abcam
AB211872-100
ABCAAB211872-100EA 638 EUR
ABCAAB211872-100
Anti-Anillin Mouse Monoclonal Antibody [clone: CL0303]
Antikörper
Mouse monoclonal [CL0303] to Anillin.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Anillin
Clonality: Monoclonal
Clone: CL0303
Conjugation:
Epitope: Binds to an epitope located within the peptide sequence LRGRFDKGNIWSAEK (438-452 aa) as determined by overlapping synthetic peptides.
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR