Anti-S100 beta Mouse Monoclonal Antibody [clone: S100B/1012]

Lieferant: Abcam
AB218956-100
ABCAAB218956-100EA 1000 EUR
ABCAAB218956-100
Anti-S100 beta Mouse Monoclonal Antibody [clone: S100B/1012]
Antikörper
Mouse monoclonal [S100B/1012] to S100 beta - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: S100 beta
Clonality: Monoclonal
Clone: S100B/1012
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR