Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB218983-50
ABCAAB218983-50EA 688 EUR
ABCAAB218983-50
Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Carbonic Anhydrase 12/CA12.

Type: Primary
Antigen: Carbonic Anhydrase 12/CA12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR