Anti-Hemoglobin subunit zeta Rabbit Polyclonal Antibody
Lieferant: Abcam
AB228651-100
ABCAAB228651-100EA 688 EUR
ABCAAB228651-100
Anti-Hemoglobin subunit zeta Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Hemoglobin subunit zeta.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Hemoglobin subunit zeta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human