Anti-Amodiaquine Mouse Monoclonal Antibody [clone: HYB 320-04]

Lieferant: Abcam
AB23487-100
ABCAAB23487-100EA 835 EUR
ABCAAB23487-100
Anti-Amodiaquine Mouse Monoclonal Antibody [clone: HYB 320-04]
Antikörper
Type: Primary
Antigen: Amodiaquine
Clonality: monoclonal
Clone: HYB 320-04
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR