Anti-Histone H3 (acetyl K56) Rabbit Monoclonal Antibody [clone: RM179]

Lieferant: Abcam
AB238307-50
ABCAAB238307-50EA 688 EUR
ABCAAB238307-50
Anti-Histone H3 (acetyl K56) Rabbit Monoclonal Antibody [clone: RM179]
Antikörper
Rabbit monoclonal [RM179] to Histone H3 (acetyl K56).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: RM179
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR