Anti-EPO-R Rabbit Monoclonal Antibody [clone: EPR22366-40]

Lieferant: Abcam
AB244202-100 AB244202-1ML
ABCAAB244202-100EA 782 EUR
ABCAAB244202-100 ABCAAB244202-1ML
Anti-EPO-R Rabbit Monoclonal Antibody [clone: EPR22366-40]
Antikörper
Rabbit monoclonal [EPR22366-40] to EPO-R.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: EPO-R
Clonality: Monoclonal
Clone: EPR22366-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR