Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB244309-100
ABCAAB244309-100EA 663 EUR
ABCAAB244309-100
Anti-Carbonic Anhydrase 12/CA12 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Carbonic Anhydrase 12/CA12.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 12/CA12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR