Anti-DDDDK tag (Binds to FLAG® tag sequence) Rabbit Polyclonal Antibody(HRP (Horseradish Peroxidase))

Lieferant: Abcam
AB2493-100
ABCAAB2493-100EA 550 EUR
ABCAAB2493-100
Anti-DDDDK tag (Binds to FLAG® tag sequence) Rabbit Polyclonal Antibody(HRP (Horseradish Peroxidase))
Antikörper
HRP Rabbit polyclonal to DDDDK tag (Binds to FLAG® tag sequence).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DDDDK tag (Binds to FLAG® tag sequence)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR