Anti-CD38 Rat Monoclonal Antibody [clone: 90] (Biotinylated)

Lieferant: Abcam
AB25004-100
ABCAAB25004-100EA 888 EUR
ABCAAB25004-100
Anti-CD38 Rat Monoclonal Antibody [clone: 90] (Biotinylated)
Antikörper
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: Monoclonal
Clone: 90
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR