Anti-CD36 Rabbit Monoclonal Antibody [clone: EPR22509-40]

Lieferant: Abcam
AB252922-100 AB252922-1ML
ABCAAB252922-100EA 800 EUR
ABCAAB252922-100 ABCAAB252922-1ML
Anti-CD36 Rabbit Monoclonal Antibody [clone: EPR22509-40]
Antikörper
Rabbit monoclonal [EPR22509-40] to CD36.

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: EPR22509-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR