Anti-Ly6g Rat Monoclonal Antibody PE (Phycoerythrin)/Cy7® [clone: RB6-8C5]

Lieferant: Abcam
AB25514-50
ABCAAB25514-50EA 843 EUR
ABCAAB25514-50
Anti-Ly6g Rat Monoclonal Antibody PE (Phycoerythrin)/Cy7® [clone: RB6-8C5]
Antikörper
Type: Primary
Antigen: Ly6g
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR