Anti-SPAM1 Rabbit Monoclonal Antibody [clone: EPR22906-57]

Lieferant: Abcam
AB256482-1 AB256482-100
ABCAAB256482-1EA 9160 EUR
ABCAAB256482-1 ABCAAB256482-100
Anti-SPAM1 Rabbit Monoclonal Antibody [clone: EPR22906-57]
Antikörper
Rabbit monoclonal [EPR22906-57] to SPAM1 - BSA and Azide free.

Type: Primary
Antigen: SPAM1
Clonality: Monoclonal
Clone: EPR22906-57
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR