Anti-HLA DR Mouse Monoclonal Antibody [clone: MEM-267]

Lieferant: Abcam
AB26089-100
ABCAAB26089-100EA 825 EUR
ABCAAB26089-100
Anti-HLA DR Mouse Monoclonal Antibody [clone: MEM-267]
Antikörper
Type: Primary
Antigen: HLA DR
Clonality: Monoclonal
Clone: MEM-267
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR