Anti-DGKZ/DGK-zeta Rabbit Monoclonal Antibody [clone: EPR22040-80] (Alexa Fluor® 647)
Lieferant: Abcam
AB270107-100
ABCAAB270107-100EA 844 EUR
ABCAAB270107-100
Anti-DGKZ/DGK-zeta Rabbit Monoclonal Antibody [clone: EPR22040-80] (Alexa Fluor® 647)
Antikörper
Alexa Fluor® 647 Rabbit monoclonal [EPR22040-80] to DGKZ/DGK-zeta.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DGKZ/DGK-zeta
Clonality: Monoclonal
Clone: EPR22040-80
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human