Anti-Carbonic Anhydrase 12/CA12 Mouse Monoclonal Antibody [clone: 11] (FITC (Fluorescein Isothiocyanate))

Lieferant: Abcam
AB275576-100
ABCAAB275576-100EA 688 EUR
ABCAAB275576-100
Anti-Carbonic Anhydrase 12/CA12 Mouse Monoclonal Antibody [clone: 11] (FITC (Fluorescein Isothiocyanate))
Antikörper
FITC Mouse monoclonal [11] to Carbonic Anhydrase 12/CA12.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Carbonic Anhydrase 12/CA12
Clonality: Monoclonal
Clone: 11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR