Anti-TLS/FUS Rabbit Monoclonal Antibody [clone: EPR5812] (APC (Allophycocyanin))
Lieferant: Abcam
AB306073-100
ABCAAB306073-100EA 713 EUR
ABCAAB306073-100
Anti-TLS/FUS Rabbit Monoclonal Antibody [clone: EPR5812] (APC (Allophycocyanin))
Antikörper
APC Rabbit monoclonal [EPR5812] to TLS/FUS.
Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TLS/FUS
Clonality: Monoclonal
Clone: EPR5812
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen