Anti-PKC delta Mouse Monoclonal Antibody [clone: 14/PKC delta]

Lieferant: Abcam
AB307233-100 AB307233-10 AB307233-20UG AB307233-1MG
ABCAAB307233-100EA 694 EUR
ABCAAB307233-100 ABCAAB307233-10 ABCAAB307233-20UG ABCAAB307233-1MG
Anti-PKC delta Mouse Monoclonal Antibody [clone: 14/PKC delta]
Antikörper
Mouse monoclonal [14/PKC delta] to PKC delta.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PKC delta
Clonality: Monoclonal
Clone: 14/PKC delta
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR