Anti-TLS/FUS Rabbit Monoclonal Antibody [clone: EPR5812] (AP (Alkaline Phosphatase))

Lieferant: Abcam
AB308861-200
ABCAAB308861-200EA 819 EUR
ABCAAB308861-200
Anti-TLS/FUS Rabbit Monoclonal Antibody [clone: EPR5812] (AP (Alkaline Phosphatase))
Antikörper
Alkaline Phosphatase Rabbit monoclonal [EPR5812] to TLS/FUS.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TLS/FUS
Clonality: Monoclonal
Clone: EPR5812
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR