Anti-PKC alpha+PKC delta+PKC epsilon+PKC beta 1+PKC gamma+PKC zeta+PKC eta+PKC theta/PRKCQ+PKC iota (phospho T410) Rabbit Monoclonal Antibody [clone: HL1279]

Lieferant: Abcam
AB313842-100UL
ABCAAB313842-100ULEA 1020 EUR
ABCAAB313842-100UL
Anti-PKC alpha+PKC delta+PKC epsilon+PKC beta 1+PKC gamma+PKC zeta+PKC eta+PKC theta/PRKCQ+PKC iota (phospho T410) Rabbit Monoclonal Antibody [clone: HL1279]
Antikörper
Rabbit monoclonal [HL1279] to PKC alpha (phospho T410) + PKC delta (phospho T410) + PKC epsilon (phospho T410) + PKC beta 1 + PKC gamma (phospho T410) + PKC zeta (phospho T410) - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PKC alpha+PKC delta+PKC epsilon+PKC beta 1+PKC gamma+PKC zeta+PKC eta+PKC theta/PRKCQ+PKC iota
Clonality: Monoclonal
Clone: HL1279
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Drosophila melanogaster
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR