Anti-PPP2R5D Mouse Monoclonal Antibody [clone: H5-D12]

Lieferant: Abcam
AB316368-100UL
ABCAAB316368-100ULEA 644 EUR
ABCAAB316368-100UL
Anti-PPP2R5D Mouse Monoclonal Antibody [clone: H5-D12]
Antikörper
Mouse monoclonal [H5-D12] to PPP2R5D.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PPP2R5D
Clonality: Monoclonal
Clone: H5-D12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR