Anti-Histone H3 (di methyl K9) Rabbit Monoclonal Antibody [clone: EP16990] (APC)

Lieferant: Abcam
AB320133-100UL
ABCAAB320133-100ULEA 819 EUR
ABCAAB320133-100UL
Anti-Histone H3 (di methyl K9) Rabbit Monoclonal Antibody [clone: EP16990] (APC)
Antikörper
APC Rabbit monoclonal [EP16990] to Histone H3 (di methyl K9) - ChIP Grade.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EP16990
Conjugation: APC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR