Anti-Tec Rabbit Monoclonal Antibody [clone: Y398]

Lieferant: Abcam
AB32368-100 AB32368-1ML
ABCAAB32368-100EA 707 EUR
ABCAAB32368-100 ABCAAB32368-1ML
Anti-Tec Rabbit Monoclonal Antibody [clone: Y398]
Antikörper
Rabbit monoclonal [Y398] to Tec.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tec
Clonality: Monoclonal
Clone: Y398
Conjugation:
Epitope: ab32368 reacts with an epitope located in the PH domain of Tec
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR