Anti-Proteasome 20S LMP2 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB3328-100
ABCAAB3328-100EA 588 EUR
ABCAAB3328-100
Anti-Proteasome 20S LMP2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to Proteasome 20S LMP2.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Proteasome 20S LMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR