Anti-TNFRSF4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MRC OX-40]

Lieferant: Abcam
AB33960-50
ABCAAB33960-50EA 344 EUR
ABCAAB33960-50
Anti-TNFRSF4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MRC OX-40]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: TNFRSF4 (tumor necrosis factor receptor superfamily, member 4)
Clonality: monoclonal
Clone: MRC OX-40
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR