Anti-DRD5 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB40656-100
ABCAAB40656-100EA 808 EUR
ABCAAB40656-100
Anti-DRD5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DRD5 (dopamine receptor D5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR