Anti-N-epsilon acetyl Lysine Mouse Monoclonal Antibody [clone: 11A1]

Lieferant: Abcam
AB409-100
ABCAAB409-100EA 939 EUR
ABCAAB409-100
Anti-N-epsilon acetyl Lysine Mouse Monoclonal Antibody [clone: 11A1]
Antikörper
Type: Primary
Antigen: N-epsilon acetyl Lysine
Clonality: monoclonal
Clone: 11A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR