Anti-FOLH1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB41034-100
ABCAAB41034-100EA 918 EUR
ABCAAB41034-100
Anti-FOLH1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR