Anti-Noradrenaline transporter Rabbit Polyclonal Antibody

Lieferant: Abcam
AB41559-50
ABCAAB41559-50EA 918 EUR
ABCAAB41559-50
Anti-Noradrenaline transporter Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Noradrenaline transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR