Anti-GABA Transporter 1 / GAT 1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB426-200
ABCAAB426-200EA 927 EUR
ABCAAB426-200
Anti-GABA Transporter 1 / GAT 1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABA Transporter 1 / GAT 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR