Anti-RNF31/HOIP Rabbit Polyclonal Antibody

Lieferant: Abcam
AB46322-100
ABCAAB46322-100EA 955 EUR
ABCAAB46322-100
Anti-RNF31/HOIP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF31/HOIP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR