Anti-4-Hydroxynonenal Rabbit Polyclonal Antibody

Lieferant: Abcam
AB46545-100
ABCAAB46545-100EA 1010 EUR
ABCAAB46545-100
Anti-4-Hydroxynonenal Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 4-Hydroxynonenal (4-hydroxynonenal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR