Anti-Plasmin Rabbit Polyclonal Antibody (Biotinylated)

Lieferant: Abcam
AB48350-150
ABCAAB48350-150EA 600 EUR
ABCAAB48350-150
Anti-Plasmin Rabbit Polyclonal Antibody (Biotinylated)
Antikörper
Biotin Rabbit polyclonal to Plasmin.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Plasmin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR